Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride Bioreagent Grade, Vion Biosciences
SCI_ED

Catalog No. S11060 Shop All Aldon Products
Change view
Click to view available options
Quantity:
10 kg
25 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S11060 10 kg
S11061 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S11060 Supplier Aldon Corporation Supplier No. VNSS700310KG
Add to Cart
Add to Cart

Bioreagent grade for molecular biology, DNase-free, Rnase-free, ≥ 99%

  • SPECIFICATIONS
  • Apperance: White Crystalline Powder
  • Purity: 99.0 - 101.5%
  • pH (5% Aqueous): 5.0 - 9.0
  • Lead: ≤ 5 ppm
  • Iron: ≤ 2 ppm
  • Insoluble Matter: ≤ 0.005%
  • Iodide: ≤ 20 ppm
  • Bromide: ≤ 100 ppm
  • Phosphate: ≤ 5 ppm
  • Sulfate: ≤ 40 ppm
  • Barium: To Pass Test
  • Calcium: ≤ 20 ppm
  • Magnesium: ≤ 10 ppm
  • Potassium: ≤ 50 ppm
  • Chlorate and Nitrate: ≤ 30 ppm
  • DNase: None Detected
  • RNase: None Detected

Chemical Identifiers

CAS 7647-14-5
Molecular Weight (g/mol) 58.45 g/mol

Specifications

Physical Form Solid
Quantity 10 kg
Percent Purity 99.0 to 101.5%
Grade Reagent
Packaging UN Plastic Pail
Chemical Name or Material Sodium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.