Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium chloride, Certified by BAM, ≥99.5%, MilliporeSigma™ Supelco™

Catalog No. 111015462 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5462 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5462 Supplier MilliporeSigma Supelco Supplier No. 7138725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 7647-14-5
Vapor Pressure 1.33 (1.00 mmHg) (at 865°C hPa)
Physical Form Neat
Linear Formula NaCl
MDL Number MFCD00003477
pH 7
Melting Point 801°C
Synonym Halite
Type Certified Reference Material
Quantity 25 g
Formula Weight 58.44
Percent Purity ≥99.5%
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.