Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride CP Granular, Macron Fine Chemicals™

Catalog No. MK7544212 Shop All Avantor Products
Change view
Click to view available options
Poly; 2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-7544-212 Poly; 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-7544-212 Supplier Avantor Supplier No. 754406
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 7647-14-5
Physical Form Granules
Quantity 2.5 kg
Molecular Formula ClNa
MDL Number MFCD00003477
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
PubChem CID 5234
ChEBI CHEBI:26710
Formula Weight 58.44
Packaging Poly
Chemical Name or Material Sodium Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.