Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, Crystal, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18613419 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
12 kg
45 kg
350 lb.
Packaging:
Glass Bottle
Poly Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-419 350 lb. Poly Bottle
18-613-413 500 g Poly Bottle
18-613-414 1 kg Poly Bottle
18-613-415 2.5 kg Glass Bottle
18-613-416 2.5 kg Poly Bottle
18-613-417 12 kg Poly Bottle
18-613-418 45 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-613-419 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1240350LBBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

pH 5.0 to 9.0
CAS Max % 1
Physical Form Crystals and/or Chunks
Quantity 350 lb.
Percent Purity 99%
Grade ACS/Reagent
Identification To Pass Test
Packaging Poly Bottle
Chemical Name or Material Sodium Chloride, Crystal

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.