Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, Granular, USP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18606415 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
250 lb.
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-415 12 kg Poly Bottle
18-606-412 6 x 500 g Poly Bottle
18-606-414 4 x 2.5 kg Poly Bottle
18-606-416 45 kg Poly Bottle
18-606-417 250 lb. Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-606-415 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO15512KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SO155, 7647-14-5, NaCl

Spectrum™ Chemical Sodium Chloride, Granular, USP is used in the formulation of oral hygiene products, shampoos, fragrance, skin, hair, nail, cleansing, suntan, makeup and bath products. This grade is specifically used in oral care applications. This product is not sterile and not intended for use in the manufacture of injectable dosage forms, peritoneal dialysis solutions, hemodialysis solutions or hemofiltration solutions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

CAS Max % 1
Physical Form Granular/Lump
Quantity 12 kg
Percent Purity 99 to 100.5%
Grade USP
Loss on Drying 0.005
Packaging Poly Bottle
Appearance Clear
Chemical Name or Material Sodium Chloride, Granular
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.