Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride Granulated, Supelco

Catalog No. M1174191000 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1174191000 1 kg
M1174195000 5 kg
M1174199025 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1174191000 Supplier MilliporeSigma Supelco Supplier No. 1174191000
Add to Cart
Add to Cart

For Analysis EMSURE™ ACS, ISO, Reag. Ph Eur

  • Experience the advantages of EMSURE™ Granulated Salts, specially designed for optimal handling and processing.
  • Unlike powdered salts, our granulated formulation offers superior flow properties, ensuring consistency in your analytical applications.
  • Our innovative dry granulation process keeps the salts free-flowing while preserving premium specifications, even during long-term storage.

Key Benefits:

  • Exceptional Purity: Achieve reliable results with EMSURE™ analytical grade salts.
  • Chemical Stability: Ensure consistent outcomes with our stable formulation.
  • Enhanced Flowability: Enjoy easier handling, improved safety, and reduced waste during use.
  • Dust-Free Handling: Benefit from a clean, dust-free experience that enhances safety and usability.
  • Unchanged Specifications: Rest assured that product specifications remain consistent and optimized for analytical applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Physical Form Granules

Specifications

CAS 7647-14-5
Quantity 1 kg
Percent Purity ≥99.5% (Argentometric), 99.0 to 100.5% (Argentometric, calculated on dried substance)
Grade ACS
Chemical Name or Material Sodium Chloride Granulated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.