Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride-Hydroxylamine Sulfate Solution, Ricca Chemical
SDP

Catalog No. 71944 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71944 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71944 Supplier Ricca Chemical Company Supplier No. 71944
Add to Cart
Add to Cart

For APHA/ASTM/EPA Testing

Specifications

Chemical Name or Material Sodium Chloride-Hydroxylamine Sulfate Solution
Packaging Natural Poly Bottle
CAS 7732-18-5, 7647-14-5, 10039-54-0
CAS Min % 79.0, 9.49, 9.49
CAS Max % 82.0, 9.88, 9.88
Physical Form Liquid
Color Colorless
Boiling Point Approximately 100°C
For Use With (Equipment) Atomic Absorption Spectrophotometer
Solubility Miscible with water
Solution Type Calibration Solution
Type Buffer Solution
Quantity 120 mL
Grade Laboratory
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.