Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sodium Chloride, Lab Grade
SCI_ED

Catalog No. S25541 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
3 kg
12 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25541 500 g
S25541A 2.5 kg
S25541B 12 kg
S25542A 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S25541
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Color White
Quantity 500 g
Assay Percent Range >95% w/v
Linear Formula NaCl
Formula Weight 58.44
Grade Laboratory
Density 2.165g/mL
Chemical Name or Material Sodium Chloride
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.