Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, MP Biomedical

Catalog No. MP210289210 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 kg
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210289210 10 kg
MP210289205 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP210289210 Supplier MP Biomedicals, Inc Supplier No. 0210289210
Add to Cart
Add to Cart

Sodium chloride, with a purity of 99.0%-100.5%, is a commonly used chemical which is found widely in nature. It is considered to be an essential nutrient. Excess amounts of sodium chloride can destroy electrolyte balance and cause death in most animals, including humans.

  • Sodium chloride is used in a wide variety of biochemical applications
  • It has also been used in high concentrations for preservation of foods since bacteria cannot grow in high salt conditions
  • Normal saline is 0.9% w/v in water
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, dendritis, purex, sodium chloric
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1461°C (1013hPa)
Melting Point 801°C (literature)
Color White
Quantity 10 kg
Percent Purity 99%
Packaging Plastic
Density 2.17 g/cm³at 20 °C (Density of a saturated solution at 25 °C is 1.202)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.