Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride, Purified, ASTM B117 Contains no anti-caking agents, Reagents

Catalog No. 01337035
Change view
Click to view available options
Quantity:
50 lb.
50 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-035 50 lb.
01-337-036 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-035 Supplier Reagents Holdings Llc Supplier No. C231310050F5
Only null left
Add to Cart
Add to Cart

Sodium Chloride, Purified, ASTM B117 Contains no anti-caking agents, ASTM, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym Table salt
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Boiling Point 1461°C
Melting Point 801°C
Color Colorless/White
Physical Form Solid
Quantity 50 lb.
Packaging Fiber Drum
Specific Gravity 2.17
Chemical Name or Material Sodium Chloride, Purified, ASTM B117 Contains no anti-caking agents
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.