Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride RV, EP Primary Standard, Ricca Chemical
SDP

Catalog No. 7227850G Shop All Ricca Chemical Company Products
Change view
Click to view available options
Package: White Poly Wide Mouth; Size: 50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
72-278-50G Package: White Poly Wide Mouth; Size: 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 72-278-50G Supplier Ricca Chemical Company Supplier No. 7227.850G
Add to Cart
Add to Cart

Specifications

Color White
CAS 7647-14-5
Physical Form Crystals
Quantity 50 g
Molecular Formula ClNa
MDL Number MFCD00003477
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
PubChem CID 5234
ChEBI CHEBI:26710
Percent Purity 100%
Grade Laboratory
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Sodium Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.