Learn More
Weber Scientific Salometer, Sodium Chloride, Plastic, Saturation
SDP
Catalog No. NC1418345
Encompass
Measures salt in water (brine). Available in sodium chloride. Standardized at 60°F. Length: 300 mm.
Catalog No. NC1418345 Supplier Weber Scientific Supplier No. 303642
Measures salt in water (brine). Available in sodium chloride. Standardized at 60°F. Length: 300 mm.