Learn More
Bioworld Sodium Chloride, 0.85-0.90% (w/v), 4 L
Catalog No. 50255587
Encompass_Preferred
Used in biochemistry and molecular biology applications; a component of PBS and SSC buffersAppearance: LiquidBoiling Point : 1413 °CDensity : 2.17 g/cm3 at 20 °CMelting Point: 801 °C (lit.)RTECS Number: VZ4725000
Catalog No. 50-255-587 Supplier Bioworld Supplier No. 402201333
Used in biochemistry and molecular biology applications; a component of PBS and SSC buffersAppearance: LiquidBoiling Point : 1413 °CDensity : 2.17 g/cm3 at 20 °CMelting Point: 801 °C (lit.)RTECS Number: VZ4725000