Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium chloride solution, 5 M in H2O, 7647-14-5, MFCD00003477, 1L
Catalog No. 501657210
Encompass
Linear Formula NaCl, Molecular Weight 58.44, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture.
Catalog No. 50-165-7210 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S51501L
Linear Formula NaCl, Molecular Weight 58.44, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture.