Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride Standard, 0.90% (w/w), Ricca Chemical
SDP

Catalog No. R721309010F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R721309010F 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R721309010F Supplier Ricca Chemical Company Supplier No. R721309010F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 7647-14-5 , 7647-14-5
Molecular Formula H2O
Molecular Weight (g/mol) 58.44 g/mol
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Melting Point 0.0°
Color Colorless
pH 6.7 to 7.3
CAS Min % 99.1%, 0.9%
CAS Max % 99.1%, 0.9%
Physical Form Liquid
Quantity 10 L
Assay Percent Range 0.895 to 0.905%
Percent Purity 0.90% (w/w)
Grade Laboratory
Packaging Cubitainer
Appearance Clear, Colorless Liquid
Assay % (w/w)
Chemical Name or Material Sodium Chloride Standard
Show More Show Less
ShelfLife 24 Months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.