Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride Standard, 100 ppm NaCl, Ricca Chemical
SDP

Catalog No. R72230001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R72230001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R72230001A Supplier Ricca Chemical Company Supplier No. R72230001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7647-14-5 , 7732-18-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
PubChem CID 5234
ChEBI CHEBI:26710
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Packaging Natural Poly Bottle
Odor Odorless
Physical Form Liquid
Color Colorless
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Analytical Standard
Quantity 1 L
Grade Laboratory
Chemical Name or Material Sodium Chloride Standard, 100 ppm NaCl
Show More Show Less
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.