Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chloride/Sulfuric Acid, Ricca Chemical
SDP

Catalog No. R72277001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R72277001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R72277001A Supplier Ricca Chemical Company Supplier No. R72277001A
Add to Cart
Add to Cart

for Method A022

Specifications

Color Colorless
CAS 7732-18-5, 7647-14-5, 7664-93-9
CAS Max % 89.39, 9.35, 1.26
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name water
Molecular Weight (g/mol) 18.02
Grade Laboratory
Packaging Natural Poly Bottle
Quantity 1 L
Chemical Name or Material Sodium Chloride-Sulfuric Acid
Show More Show Less
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.