Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium Chloride Tablets - CAS 7647-14-5 - Calbiochem Each tablet yields 1 L of 0.9% sodium chloride. | 7647-14-5 | MFCD00003477 | 1EA
Catalog No. 501752201
Encompass
Sodium Chloride Tablets - CAS 7647-14-5 - Calbiochem Each tablet yields 1 L of 0.9% sodium chloride. | Mol Wt: 58.44 | 7647-14-5 | MFCD00003477 | 1EA
Catalog No. 50-175-2201 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 5674421EA
Sodium Chloride Tablets - CAS 7647-14-5 - Calbiochem Each tablet yields 1 L of 0.9% sodium chloride. | Mol Wt: 58.44 | 7647-14-5 | MFCD00003477 | 1EA