Learn More
Chemical Services Laboratories Sodium Chloride for standardizing 0.01 N Silver Nitrate | 7647-14-5 | MFCD00003477 | 12 ampules/box
Catalog No. NC9777815
Encompass
Sodium Chloride for standardizing 0.01 N Silver Nitrate | MW: 58.443 | 7647-14-5 | MFCD00003477 | 12 ampules/box
Catalog No. NC9777815 Supplier Chemical Services Laboratories Supplier No. 1004
Add to Cart
Add to Cart
Sodium Chloride for standardizing 0.01 N Silver Nitrate | MW: 58.443 | 7647-14-5 | MFCD00003477 | 12 ampules/box