Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STE Buffer, Sodium Chloride-Tris-EDTA, 10X Solution, pH 8.0, Fisher BioReagents

Catalog No. BP24791 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP24791 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP24791

missing translation for 'validMainframeMsg'

  • Save time and simplify your buffer preparation step by using Fisher BioReagents 10X STE buffer solution - simply dilute as needed
  • When diluted, 1X solution contains 10mM tris, 1mM EDTA, and 100mM NaCl with pH 8.0 at 25°C
  • Filtered through a 5 micron filter and autoclaved
  • Tested for the absence of DNase, RNase, and protease
  • STE buffer is used for DNA extraction protocols
  • Intended for laboratory research use only
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Chloride-Tris-EDTA (STE) Buffer
Name Note 10X, pH 8.0
CAS 7647-14-5, 60-00-4, 7647-01-0, 7732-18-5
Boiling Point 100°C
Color Undesignated
pH 8
Physical Form Liquid
Quantity 1 L
DNase DNase free
Filtered Through Filtered through a 5-micron filter.
Formulation Water:>90% Sodium Chloride: <10% 1,3-Propanediol, 2-amino-2-(hydroxymethyl)-hydrochloride: <5% Ethylenediamine tetraacetic acid (EDTA): <1.0%
Packaging Nalgene Poly Bottle
Protease Protease free
Synonym Saline Tris–EDTA, STE, TEN
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.