Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Sodium Chloride USP
SDP

Catalog No. 14186089 Shop All Medisca Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
25 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-186-089 25 kg
14-185-255 500 g
14-185-556 1 kg
14-185-667 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-186-089 Supplier Medisca Supplier No. 062907
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Chloride USP can be colorless, cubic crystals or white, crystalline powder. Alternative name(s): salt

Sodium Chloride USP can be colorless, cubic crystals or white, crystalline powder.

Alternative name(s): salt

Specifications

Chemical Name or Material Sodium Chloride USP
Packaging Pail, 6 gallon, White, HDPE, Round
CAS 7647-14-5
Type Drug Standard
Molecular Weight (g/mol) 58.443
Quantity 25 kg
Percent Purity 99 to 100.5 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.