Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Chlorite, Anhydrous, Flakes, 77.5-82.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
Description
Spectrum™ Chemical Sodium Chlorite, Anhydrous, Flakes is a chemical compound often used in the manufacture of paper. Used to generate chlorine dioxide for the bleaching and striping of textiles it is also frequently used as a oxidation reagent in organic synthesis. (CAS No. 7758-19-2, Molecular Formula: NaClO2) Sodium Chlorite, Anhydrous, Flakes
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 2.5 kg |
| Percent Purity | 77.5 to 82.5% |
| Grade | Reagent |
| Packaging | Poly Pail |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction