Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chlorite, Anhydrous, Flakes, 77.5-82.5%, Spectrum™ Chemical
SDP

Catalog No. 18606435 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-435 12 kg Poly Pail
18-606-436 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-606-435 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S124312KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

S1243, 7758-19-2, NaClO2

Spectrum™ Chemical Sodium Chlorite, Anhydrous, Flakes is a chemical compound often used in the manufacture of paper. Used to generate chlorine dioxide for the bleaching and striping of textiles it is also frequently used as a oxidation reagent in organic synthesis.

(CAS No. 7758-19-2, Molecular Formula: NaClO2) Sodium Chlorite, Anhydrous, Flakes

Chemical Identifiers

CAS 7758-19-2
Molecular Formula ClNaO2
Molecular Weight (g/mol) 90.44
InChI Key UKLNMMHNWFDKNT-UHFFFAOYSA-M
IUPAC Name sodium chlorite
SMILES [Na+].[O-][Cl]=O

Specifications

CAS Max % 1
Quantity 12 kg
Percent Purity 77.5 to 82.5%
Grade Reagent
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.