Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chlorite, puriss p.a, 80% (RT), Solstice

Catalog No. 60047363 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-363 1 kg Poly bottle
60-047-364 250 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-363 Supplier Solstice Supplier No. 713881KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., 80% (RT)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-19-2
Molecular Formula ClNaO2
Molecular Weight (g/mol) 90.438
MDL Number MFCD00003478
InChI Key UKLNMMHNWFDKNT-UHFFFAOYSA-M
Synonym sodium chlorite, chlorous acid, sodium salt, textone, chlorite sodium, textile, alcide ld, neo silox d, caswell no. 755, sodiumchlorite, unii-g538ebv4vf
PubChem CID 23668197
ChEBI CHEBI:78667
IUPAC Name sodium;chlorite
SMILES [O-]Cl=O.[Na+]

Specifications

Melting Point 180°C
Color White
pH 10 to 11 (at 20°C)
Physical Form Solid
Quantity 1 kg
Packaging Poly bottle
Linear Formula NaClO2
UN Number UN1496
Solubility Information 572g/L (at 20°C , pH 7.0)
Formula Weight 58.44g/mol
Percent Purity 80% (RT)
Grade puriss. p.a.
Chemical Name or Material Sodium chlorite
Show More Show Less
Hazard Statement Missing Phrase - N15.00950417-P210-P280-P303 + P361 + P353-P305 + P351 + P338-P370 + P378
DOTInformation 5.1 / PGII
RTECSNumber 231-836-6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.