Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sodium Chromate Anhydrous
SCI_ED

Catalog No. S25544 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Lab Grade
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25544 100 g Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25544

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 7775-11-3
Molecular Formula CrNa2O4
Molecular Weight (g/mol) 161.97
InChI Key PXLIDIMHPNPGMH-UHFFFAOYSA-N
Synonym Disodium chromate, Chromium disodium oxide, Chromium sodium oxide
PubChem CID 24488
ChEBI CHEBI:65242
IUPAC Name disodium dioxochromiumbis(olate)
SMILES [Na+].[Na+].[O-][Cr]([O-])(=O)=O

Specifications

Quantity 100 g
Assay Percent Range 1
Linear Formula Na2CrO4
Formula Weight 161.97
Grade Laboratory
Packaging Lab Grade
Chemical Name or Material Sodium Chromate, Anhydrous
Safety handling Check Alert storage code Red
ShelfLife 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.