Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chromate, Anhydrous, Reagents

Catalog No. 01337034
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-034 100 g
01-337-033 3 kg
01-337-032 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-034 Supplier Reagents Holdings Llc Supplier No. C2314000100C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Chromate, Anhydrous, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7775-11-3
Molecular Formula CrNa2O4
Molecular Weight (g/mol) 161.97
InChI Key PXLIDIMHPNPGMH-UHFFFAOYSA-N
Synonym Chromium disodium oxide
IUPAC Name disodium dioxochromiumbis(olate)
SMILES [Na+].[Na+].[O-][Cr]([O-])(=O)=O

Specifications

Melting Point 792°C
Color Yellow
Physical Form Solid
Quantity 100 g
Grade Reagent
Packaging Glass Bottle
Specific Gravity 2.723
Chemical Name or Material Sodium Chromate, Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.