Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Chromate Indicator, Reagents

Catalog No. 01333865
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-865 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-865 Supplier Reagents Holdings Llc Supplier No. CS102900500A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Chromate Indicator, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium Chromate Indicator
CAS 7732-18-5, 10034-82-9
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.103
Molecular Formula Na2CrO4/H2O
Color Yellow
Synonym Chromium disodium oxide
pH 10
Quantity 500 mL
Physical Form Liquid
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.