Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Chromate Tetrahydrate

Catalog No. 216623500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-662-3500G 500 g
21-662-3100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Danger
Catalog No. 21-662-3500G Supplier Sigma Aldrich Supplier No. 216623500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium chromate tetrahydrate can be used as an oxidant in organic synthesis and also as a carrier electrolyte in the capillary electrophoresis.

Specifications

CAS 10034-82-9
Molecular Formula CrNa2O4 -+ 4H2O
Linear Formula Na2CrO4 -+ 4H2O
MDL Number MFCD00149167
Quantity 500 g
Molecular Weight (g/mol) 234.03 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.