Learn More
Thermo Scientific Chemicals Sodium citrate, 0.5 M Buffer Solution pH 6.0 PROMO
Description
- Applications: Sodium citrate, 0.5 M buffer solution, pH 6.0 is used for RNA isolation. It aids in the detection of antigens in fixed tissue preparations.
- Solubility: Fully miscible in water.
- Notes: Incompatible with heat and oxidizing agents.
Specifications
Specifications
| Chemical Name or Material | Sodium citrate Buffer Solution |
| Color | Colorless |
| Concentration | 0.5 M |
| pH | 6.0 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Odor | Odorless |
| MDL Number | MFCD00012462 |
| Solubility Information | Fully miscible in water. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.