Learn More
Boston Bioproducts Inc Citrate Buffer (0.5 M, pH 3.0) - 250ml
Catalog No. NC1711669
Encompass
The Citrate Buffer comprising of sodium citrate dihydrate and citric acid is commonly used to maintain pH levels within the range of 3.0 to 6.2. This buffer supports several applications in molecular biology, pathology, histology, immunohistochemistry, and RNA isolation. It also aids in the detection of antigens in fixed tissue preparations as it induces the breaking cross-links between antigens and components in the fixation medium.
Catalog No. NC1711669 Supplier Boston Bioproducts Inc Supplier No. BB2028250ML
The Citrate Buffer comprising of sodium citrate dihydrate and citric acid is commonly used to maintain pH levels within the range of 3.0 to 6.2. This buffer supports several applications in molecular biology, pathology, histology, immunohistochemistry, and RNA isolation. It also aids in the detection of antigens in fixed tissue preparations as it induces the breaking cross-links between antigens and components in the fixation medium.