Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Citrate, 10% w/w (10 g and 90 mL Water), Reagents

Catalog No. 01333738
Change view
Click to view available options
Quantity: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-333-738 Quantity: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-738 Supplier Reagents Holdings Llc Supplier No. CS129800500A
 Restricted Item
Add to Cart
Add to Cart

Sodium Citrate, 10% w/w (10 g + 90 mL Water), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Trisodium Citrate
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
pH 8 to 8.4
Physical Form Liquid
Quantity 500 mL
Percent Purity 10%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0435
Chemical Name or Material Sodium Citrate, 10% w/w
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.