Learn More
Med Vet International Sodium Citrate 4% 250mL Lot, J0521
Catalog No. 5000582226
Encompass_Preferred
250ml. Great for use in equine plasma therapy or small animal short-term whole blood therapy. Dilute 1ml to 7-9ml of blood
Catalog No. 50-005-82226 Supplier Med Vet International Supplier No. JOR521
250ml. Great for use in equine plasma therapy or small animal short-term whole blood therapy. Dilute 1ml to 7-9ml of blood