Learn More
Medchemexpress LLC Sodium Citrate Buffer | 50 ML
SDP
Catalog No. 5000413038
Encompass_Preferred
Sodium citrate buffer, 0.5M, pH 6.0, is a commonly used buffer. Its main components are citric acid and sodium hydrogen phosphate. It is used for RNA isolation and antigen detection in fixed tissue.
  • Commonly used buffer
  • Main components are citric acid and sodium hydrogen phosphate
  • Used for RNA isolation
  • Used for antigen detection in fixed tissue
  • Liquid appearance
  • Colorless to light yellow
Catalog No. 50-004-13038 Supplier Medchemexpress LLC Supplier No. HYB1610L50ML
Sodium citrate buffer, 0.5M, pH 6.0, is a commonly used buffer. Its main components are citric acid and sodium hydrogen phosphate. It is used for RNA isolation and antigen detection in fixed tissue.
  • Commonly used buffer
  • Main components are citric acid and sodium hydrogen phosphate
  • Used for RNA isolation
  • Used for antigen detection in fixed tissue
  • Liquid appearance
  • Colorless to light yellow