Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Citrate Dibasic Sesquihydrate

Catalog No. 71635250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-635-250G 250 g
71-635-1KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-635-250G Supplier Sigma Aldrich Supplier No. 71635250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium citrate dibasic sesquihydrate is a colorless crystalline sodium salt.Sodium citrate dibasic sesquihydrate may be employed in the following studies:In the Supel Que Citrate (EN) tubes for the quantification of pesticides in blackcurrants by triple quadrupole GC-QqQ-MS/MS [Gas chromatography coupled to tandem mass spectrometry (GC-MS/MS) coupled with triple quadrupole (QqQ) analyzer] method. Dispersive solid phase extraction (dSPE) method, commonly referred as QuEChERS.2Preparation of RNA precipitation solution.

Specifications

CAS 6132-05-4
Molecular Formula C6H6Na2O7 -+ 1.5H2O
Linear Formula HOC(COOH)(CH2COONa)2 -+ 1.5H2O
Quantity 250 g
Synonym Citric acid disodium salt; Disodium hydrogen citrate sesquihydrate
Molecular Weight (g/mol) 263.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.