Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Citrate Dihydrate, ACS Reagent Trisodium Citrate Dihydrate, Reagents

Catalog No. 01337737
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-737 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-737 Supplier Reagents Holdings Llc Supplier No. C1314500500A5
Add to Cart
Add to Cart

Sodium Citrate Dihydrate, ACS Reagent Trisodium Citrate Dihydrate, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 4-3-6132
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.10
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Synonym Trisodium Citrate Dihydrate, 2-Hydroxy-1,2,3-propanetricarboxylic Acid Trisodium Salt Dihydrate
IUPAC Name trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
SMILES O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Melting Point >300°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade ACS
Packaging Poly Bottle
Specific Gravity 1.665
Chemical Name or Material Sodium Citrate Dihydrate, ACS Reagent Trisodium Citrate Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.