Learn More
Hach Company Sodium Citrate Solution, 500 mL
Catalog No. 501995147 Shop All Hach Company Products
Encompass
For phosphate interference removal in silica tests. 500 mL bottle.
Catalog No. 50-199-5147 Supplier Hach Company Supplier No. 1490849
For phosphate interference removal in silica tests. 500 mL bottle.