Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium citrate monobasic

Catalog No. 714981KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-498-1KG 1 kg
71-498-250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-498-1KG Supplier Sigma Aldrich Supplier No. 714981KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Sodium citrate monobasic (sodium dihydrogen citrate) is the monosodium salt of citric acid
  • Isopiestic method has been proposed for the evaluation of vapor-liquid equilibrium parameters (water activity, vapor pressure, osmotic coefficient and activity coefficient) for the binary aqueous mixtures of sodium dihydrogen citrate
  • X-ray diffraction method has been reported to investigate the crystal structure of sodium dihydrogen citrate
  • Its crystals are monoclinic having space group of p21/a.sodium citrate monobasic is suitable for use in the synthesis of 16 functional alkene-terminated hyperbranched oligomer, useful for the preparation of composite system for dental composite applications
  • It is suitable for use in the preparation of gastric solutions, to investigate the effects of cooking methods and of the co-ingestion of selected food items on hg bioaccessibility in fish species

Specifications

CAS 18996-35-5
Molecular Formula C6H7NaO7
Linear Formula HOC(COONa)(CH2COOH)2
MDL Number MFCD00013067
Quantity 1 kg
Synonym Citric acid monosodium salt; Sodium dihydrogen citrate
Molecular Weight (g/mol) 214.11
Percent Purity ≥99.0% (T)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.