Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Citrate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014888 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4888 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4888 Supplier MilliporeSigma Supelco Supplier No. PHR14161G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium citrate, also known as E331, is a tri-sodium salt of citric acid. In food products, it is commonly used as a flavor and preservative. In pharmaceutical formulations, it is used as a buffer, emulsifier and sequestering agent.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
TRUSTED_SUSTAINABILITY

Specifications

CAS 4-3-6132
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Physical Form Neat
Linear Formula HOC(COONa)(CH2 COONa)2 · 2H2 O
MDL Number MFCD00150031
Melting Point >300°C
Synonym Sodium citrate tribasic dihydrate; Citric acid trisodium salt dihydrate; Trisodium citrate dihydrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 294.10
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.