Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Citrate Tribasic Dihydrate

Catalog No. S46415KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S46415KG 5 kg
S4641500G 500 g
S464125G 25 g
S46411KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S46415KG Supplier Sigma Aldrich Supplier No. S46415KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium Citrate Tribasic Dihydrate

Specifications

CAS 7681-55-2
Melting Point >300°C
Molecular Formula C6H5Na3O7 -+ 2H2O
Linear Formula HOC(COONa)(CH2COONa)2 -+ 2H2O
Quantity 5 kg
MDL Number MFCD00150031
Synonym Citric acid trisodium salt dihydrate; Trisodium citrate dihydrate
Molecular Weight (g/mol) 294.1 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.