Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Sodium Citrate,Dihydrate, OmniPur™, Calbiochem™,

Catalog No. 78605KG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 kg
Packaging:
Plastic Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
78-605-KG 5 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 78-605-KG Supplier MilliporeSigma™ Supplier No. 78605KG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5
Molecular Formula Na3C6H5O7·2H2O
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Color White
Physical Form Crystalline Powder
Quantity 5 kg
Percent Purity ≥99.0% (dried basis)
Packaging Plastic Drum
Chemical Name or Material Sodium Citrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.