Learn More
Pickering Laboratories Inc SODIUM COLUMN 4X150MM
SDP
Catalog No. NC9406469
Encompass_Preferred
High-efficiency Sodium Cation-exchange Column for protein and collegen hydrolysates, 4.0 x 150 mm, includes Amino Acid Test Mixture 1700-0070 Amino Acid Analysis.
Catalog No. NC9406469 Supplier Pickering Laboratories Inc Supplier No. 1154150
High-efficiency Sodium Cation-exchange Column for protein and collegen hydrolysates, 4.0 x 150 mm, includes Amino Acid Test Mixture 1700-0070 Amino Acid Analysis.