Learn More
Medchemexpress LLC Sodium copper chlorophyllin B | 28302-36-5 | 10 mM * 1 mL
SDP
Catalog No. 5000411005
Encompass_Preferred
Sodium copper chlorophyllin B is a research chemical known for its antiviral activities against Influenza virus and HIV. It is supplied as a solid, black-green to black in color, and has a molecular weight of 738.13. This chemical is soluble in H2O and DMSO with the aid of ultrasonication.
  • Exerts antiviral activities against Influenza virus and HIV
  • Inhibits influenza virus adsorption at 200 μM
  • Inhibits HIV adsorption at 2.5 mM
  • Soluble in H2O and DMSO
Catalog No. 50-004-11005 Supplier Medchemexpress LLC Supplier No. HYB222610MM/1ML
Sodium copper chlorophyllin B is a research chemical known for its antiviral activities against Influenza virus and HIV. It is supplied as a solid, black-green to black in color, and has a molecular weight of 738.13. This chemical is soluble in H2O and DMSO with the aid of ultrasonication.
  • Exerts antiviral activities against Influenza virus and HIV
  • Inhibits influenza virus adsorption at 200 μM
  • Inhibits HIV adsorption at 2.5 mM
  • Soluble in H2O and DMSO