Learn More
Medchemexpress LLC Sodium copper chlorophyllin B | 28302-36-5 | 500 MG
SDP
Catalog No. 5000410258
Encompass_Preferred
Sodium copper chlorophyllin B is a compound that demonstrates antiviral activities against Influenza virus and HIV, with IC50s ranging from 50 to 100 μM for both. It is intended for research use only and exhibits inhibition of influenza virus adsorption at 200 μM and HIV adsorption at 2.5 mM.
  • Antiviral activity against influenza virus and HIV.
  • IC50s of 50 to 100 μM for both influenza virus and HIV.
  • Inhibits influenza virus adsorption at 200 μM.
  • Inhibits HIV adsorption at 2.5 mM.
  • Intended for research use only.
Catalog No. 50-004-10258 Supplier Medchemexpress LLC Supplier No. HYB2226500MG
Sodium copper chlorophyllin B is a compound that demonstrates antiviral activities against Influenza virus and HIV, with IC50s ranging from 50 to 100 μM for both. It is intended for research use only and exhibits inhibition of influenza virus adsorption at 200 μM and HIV adsorption at 2.5 mM.
  • Antiviral activity against influenza virus and HIV.
  • IC50s of 50 to 100 μM for both influenza virus and HIV.
  • Inhibits influenza virus adsorption at 200 μM.
  • Inhibits HIV adsorption at 2.5 mM.
  • Intended for research use only.