Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium cyanoborohydride, 1M solution in THF, AcroSeal™

Catalog No. p-3758762
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC372451000 100 mL AcroSeal™ Glass bottle
AC372458000 800 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC372451000 Supplier Thermo Scientific Chemicals Supplier No. 372451000
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 25895-60-7 | CH3BNNa | 62.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium cyanoborohydride
Name Note 1M solution in THF
CAS 25895-60-7, 109-99-9
Quantity 100 mL
Molecular Formula CH3BNNa
Linear Formula NaBH3CN
MDL Number MFCD00003516
Flash Point −18°C
Merck Index 15, 8742
Synonym sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1-
Solubility Information Solubility in water: reacts. Other solubilities: soluble in methanol; soluble in thf, 372g/L, at 28°C, 410g/L at 46°C, 422g/L at 62°C, soluble in diglyme 176g/L at 25°C, slightly soluble in ethanol, insoluble in ether, benzene, hexane
InChI Key CVDUGUOQTVTBJH-UHFFFAOYSA-N
SMILES [Na+].[BH3-]C#N
IUPAC Name sodium;cyanoboron(1-)
Molecular Weight (g/mol) 62.84
PubChem CID 20587905
Formula Weight 62.84
Packaging AcroSeal™ Glass bottle
Appearance Clear colorless solution
Specific Gravity 0.915
Density 0.9150g/mL
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.