Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Cyanoborohydride Solution

Catalog No. 29694550ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-694-550ML 50 mL
29-694-5500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 29-694-550ML Supplier Sigma Aldrich Supplier No. 29694550ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium cyanoborohydride is a selective, mild reducing reagent for a number of functional groups such as aldehydes, ketones, acetals, epoxides and imines.Reactant for:Direct conversion of heterocyclic aldehydes to esters (oxidizing behavior)Reductive amination reactionsIntramolecular reductive cyclization reactionsReductions reactionsPreparation of dicyanoborate-based ionic liquids as hypergolic fluids for bIpr∗

Specifications

CAS 25895-60-7
Density 1.083 g/mL at 25°C
Molecular Formula CH3BNNa
Linear Formula NaBH3CN
MDL Number MFCD00003516
Quantity 50 mL
Synonym Sodium borocyanohydride; Sodium cyanoborohydride; Sodium cyanotrihydroborate
Molecular Weight (g/mol) 62.84 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.