Learn More
Medchemexpress LLC Sodium cyclamate | 139-05-9 | MFCD00003827 | 99.9% | 201.22 | C6H12NNaO3S | 500 MG
SDP
Catalog No. 5000302684
Encompass_Preferred
Sodium cyclamate analytical standard (CAS 139-05-9) is a high-purity reference material intended for analytical and research use. Provided as a stable crystalline powder, it supports method development, calibration, and quality control in analytical laboratories.
  • High purity suitable for reference and calibration.
  • Molecular formula C6H12NNaO3S; molecular weight 201.22.
  • Intended for analytical method development and validation.
  • Supplied as a stable white crystalline powder for easy handling.
  • Compatible with chromatographic and spectrometric analysis.
Catalog No. 50-003-02684 Supplier Medchemexpress LLC Supplier No. HYW014839R500MG
Sodium cyclamate analytical standard (CAS 139-05-9) is a high-purity reference material intended for analytical and research use. Provided as a stable crystalline powder, it supports method development, calibration, and quality control in analytical laboratories.
  • High purity suitable for reference and calibration.
  • Molecular formula C6H12NNaO3S; molecular weight 201.22.
  • Intended for analytical method development and validation.
  • Supplied as a stable white crystalline powder for easy handling.
  • Compatible with chromatographic and spectrometric analysis.