Learn More
Medchemexpress LLC Sodium cyclamate | 139-05-9 | MFCD00003827 | 99.9% | 201.22 g/mol | C6H12NNaO3S | 250 MG
SDP
Catalog No. 5000302781
Encompass_Preferred
Cyclamic acid sodium (sodium cyclamate) is the sodium salt of cyclamic acid supplied as a high-purity analytical standard for research and laboratory use. It is intended as a reference material for chromatographic analysis, method validation, and quantitative testing.
  • High purity (99.94%) suitable for analytical applications.
  • Available in multiple pack sizes, including 250 mg, for flexible use.
  • Molecular weight 201.22 g/mol; chemical formula C6H12NNaO3S.
  • Stable, crystalline powder form for straightforward handling and storage.
  • Intended uses include reference standard, calibration, and method development.
Catalog No. 50-003-02781 Supplier Medchemexpress LLC Supplier No. 250MGHYW014839R
Cyclamic acid sodium (sodium cyclamate) is the sodium salt of cyclamic acid supplied as a high-purity analytical standard for research and laboratory use. It is intended as a reference material for chromatographic analysis, method validation, and quantitative testing.
  • High purity (99.94%) suitable for analytical applications.
  • Available in multiple pack sizes, including 250 mg, for flexible use.
  • Molecular weight 201.22 g/mol; chemical formula C6H12NNaO3S.
  • Stable, crystalline powder form for straightforward handling and storage.
  • Intended uses include reference standard, calibration, and method development.