Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Sodium D-glycero-D-guloheptonate dihydrate, 98+%

Catalog No. AAH3670322
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3670322 100 g
AAH3670336 500 g
AAH36703A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3670322 Supplier Thermo Scientific™ Supplier No. H3670322
Add to Cart

CAS: 13007-85-7 | C7H13NaO8 | 248.16 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13007-85-7
Molecular Formula C7H13NaO8
Molecular Weight (g/mol) 248.16
InChI Key FMYOMWCQJXWGEN-WYRLRVFGSA-M
Synonym D-Glucoheptonic acid sodium salt dihydrate;
IUPAC Name sodium (2R,3R,4S,5R,6R)-2,3,4,5,6,7-hexahydroxyheptanoate
SMILES [Na+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)[C@@H](O)C([O-])=O

Specifications

Physical Form Solid
Quantity 100 g
Formula Weight 284.19
Percent Purity ≥99%
ShelfLife 5 Years
EINECSNumber 235-849-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.