Learn More
Medchemexpress LLC Sodium decane-1-sulfonate | 13419-61-9 | 244.33 | 1 G
SDP
Catalog No. 5000365382
Encompass_Preferred
Sodium decane-1-sulfonate is an anionic surfactant primarily used for the analysis of organic small molecule compounds by HPLC and ion-pair liquid chromatography. It appears as a solid and is intended for research use only.
  • Anionic surfactant
  • Used for analysis of organic small molecule compounds
  • Applicable in HPLC and ion-pair liquid chromatography
  • Solid appearance
  • For research use only
Catalog No. 50-003-65382 Supplier Medchemexpress LLC Supplier No. 1GHYW009545
Sodium decane-1-sulfonate is an anionic surfactant primarily used for the analysis of organic small molecule compounds by HPLC and ion-pair liquid chromatography. It appears as a solid and is intended for research use only.
  • Anionic surfactant
  • Used for analysis of organic small molecule compounds
  • Applicable in HPLC and ion-pair liquid chromatography
  • Solid appearance
  • For research use only