Learn More
Medchemexpress LLC Sodium decane-1-sulfonate | 13419-61-9 | 244.33 | 5 G
SDP
Catalog No. 5000364479
Encompass_Preferred
Sodium decane-1-sulfonate is an anionic surfactant primarily used for the analysis of organic small molecule compounds via HPLC and ion-pair liquid chromatography. This product is intended for research use only and is not sold to patients.
  • Anionic surfactant
  • Used for analysis of organic small molecule compounds
  • Applied in HPLC
  • Applied in ion-pair liquid chromatography
  • For research use only
Catalog No. 50-003-64479 Supplier Medchemexpress LLC Supplier No. 5GHYW009545
Sodium decane-1-sulfonate is an anionic surfactant primarily used for the analysis of organic small molecule compounds via HPLC and ion-pair liquid chromatography. This product is intended for research use only and is not sold to patients.
  • Anionic surfactant
  • Used for analysis of organic small molecule compounds
  • Applied in HPLC
  • Applied in ion-pair liquid chromatography
  • For research use only